siRNA Details
| Gene Symbol: TIMM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004085.3 | NP_004076.1 | 2 | 1255 | ||
| GenBank mRNA | BC006994.1 | - | 2 | 1005 | 4051368 | |
| BC015093.1 | - | 2 | 1008 | 3684305 | ||
| BC070284.1 | - | 2 | 988 | 4281013 | ||
| U66035.1 | - | 2 | 985 | |||

TIMM8A
Genomic Map