siRNA Details
| Gene Symbol: SNAPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003082.3 | NP_003073.1 | 10 | 1737 | ||
| GenBank mRNA | BC014984.1 | - | 10 | 1645 | 4872279 | |
| BC019038.2 | - | 10 | 1641 | 4590014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAPC1| Gene Symbol: SNAPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003082.3 | NP_003073.1 | 10 | 1737 | ||
| GenBank mRNA | BC014984.1 | - | 10 | 1645 | 4872279 | |
| BC019038.2 | - | 10 | 1641 | 4590014 | ||