siRNA Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003137.4 | NP_003128.3 | 5 | 493 | ||
| NR_034069.1 | 5 | 521 | ||||
| GenBank mRNA | AJ318054.1 | - | 4 | 891 | ||
| AK299591.1 | - | 5 | 481 | |||
| AK312712.1 | - | 5 | 385 | |||
| BC038292.1 | - | 5 | 458 | 4824261 | ||
| BG772464.1 | - | 5 | 501 | |||
| BX648663.1 | - | 5 | 410 | |||
| BX648697.1 | - | 5 | 411 | |||
| DB233331.1 | - | 5 | 493 | |||

SRPK1
Genomic Map