siRNA Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003137.4 | NP_003128.3 | 10 | 1092 | ||
| NR_034069.1 | 10 | 1120 | ||||
| GenBank mRNA | AJ318054.1 | - | 9 | 1490 | ||
| AK292277.1 | - | 10 | 1037 | |||
| AK299591.1 | - | 10 | 1080 | |||
| AK312712.1 | - | 10 | 984 | |||
| BC038292.1 | - | 10 | 1057 | 4824261 | ||
| BX648663.1 | - | 10 | 1009 | |||
| BX648697.1 | - | 10 | 1010 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016795, AJ224115, AK140761, AK161346, AB012290, and others.

SRPK1
Genomic Map