siRNA Details
| Gene Symbol: THAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004705.3 | NP_004696.2 | 5 | 1121 | ||
| NR_130898.1 | 6 | 1216 | ||||
| GenBank mRNA | AF007393.1 | - | 5 | 992 | ||
| AF081567.1 | - | 5 | 992 | |||
| AK291843.1 | - | 5 | 993 | |||
| AK299594.1 | - | 3 | 752 | |||
| AL049970.1 | - | 1 | 524 | |||
| BC110304.1 | - | 5 | 1098 | 5222582 | ||
| BC117138.1 | - | 5 | 1050 | 40125689 | ||
| BC117140.1 | - | 5 | 1050 | 40125691 | ||
| BX641144.1 | - | 2 | 4592 | |||

THAP12
Genomic Map