siRNA Details
| Gene Symbol: THAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004705.3 | NP_004696.2 | 5 | 2829 | ||
| NR_130898.1 | 6 | 2924 | ||||
| GenBank mRNA | AF081567.1 | - | 5 | 2700 | ||
| AK001522.1 | - | 1 | 1268 | |||
| AK291843.1 | - | 5 | 2701 | |||
| AL049970.1 | - | 1 | 2233 | |||
| BC021992.1 | - | 1 | 1119 | |||
| BC110304.1 | - | 5 | 2806 | 5222582 | ||
| BX641144.1 | - | 2 | 6300 | |||

THAP12
Genomic Map