siRNA Details
Gene Symbol: PRKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005044.4 | NP_005035.1 | 9 | 4111 | ||
GenBank mRNA | X85545.1 | - | 9 | 4085 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005044.4 | NP_005035.1 | 9 | 4111 | ||
GenBank mRNA | X85545.1 | - | 9 | 4085 |