siRNA Details
| Gene Symbol: PWP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005049.2 | NP_005040.2 | 21 | 3127 | ||
| GenBank mRNA | BC013309.2 | - | 21 | 3037 | 3528790 | |
| BC014988.1 | - | 21 | 3037 | 4906611 | ||
| U53346.1 | - | 21 | 3061 | |||
| U56085.1 | - | 21 | 3072 | |||
| X95263.1 | - | 21 | 3068 | |||
| Gene Symbol: LOC102724159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006723910.1 | XP_006723973.1 | 21 | 3164 | ||

PWP2
Genomic Map