siRNA Details
| Gene Symbol: CXCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302123.1 | NP_001289052.1 | 4 | 651 | ||
| NM_005409.4 | NP_005400.1 | 4 | 655 | |||
| GenBank mRNA | AF002985.1 | - | 4 | 532 | ||
| AF030514.1 | - | 4 | 514 | |||
| BC005292.1 | - | 4 | 505 | 3934139 | ||
| BC012532.1 | - | 4 | 659 | 4096572 | ||
| BC110986.1 | - | 4 | 514 | 5178174 | ||
| U66096.1 | - | 4 | 490 | |||

CXCL11
Genomic Map