siRNA Details
| Gene Symbol: SS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007559.2 | NP_001007560.1 | 5 | 666 | ||
| NM_001308201.1 | NP_001295130.1 | 5 | 641 | |||
| NM_005637.3 | NP_005628.2 | 5 | 666 | |||
| XM_006722527.2 | XP_006722590.1 | 6 | 692 | |||
| XM_011526145.1 | XP_011524447.1 | 5 | 634 | |||
| XM_011526147.2 | XP_011524449.1 | 7 | 917 | |||
| XM_011526148.2 | XP_011524450.1 | 7 | 904 | |||
| XM_011526149.2 | XP_011524451.1 | 7 | 929 | |||
| XM_011526150.2 | XP_011524452.1 | 6 | 789 | |||
| XM_011526151.2 | XP_011524453.1 | 7 | 918 | |||
| XM_011526152.2 | XP_011524454.1 | 6 | 789 | |||
| GenBank mRNA | AF244972.1 | - | 5 | 603 | ||
| AF343880.1 | - | 5 | 600 | |||
| AK289510.1 | - | 5 | 644 | |||
| AK296949.1 | - | 5 | 624 | |||
| AK299082.1 | - | 4 | 534 | |||
| AK301504.1 | - | 5 | 671 | |||
| AK304305.1 | - | 6 | 782 | |||
| AK307923.1 | - | 4 | 557 | |||
| AK316302.1 | - | 5 | 616 | |||
| AK316338.1 | - | 6 | 735 | |||
| BC096221.1 | - | 5 | 603 | 40004908 | ||
| BC096222.1 | - | 5 | 603 | 40004909 | ||
| BC096223.1 | - | 5 | 603 | 40004911 | ||
| BC096224.1 | - | 5 | 603 | 40004912 | ||
| CR542088.1 | - | 5 | 600 | |||
| CR542103.1 | - | 5 | 600 | |||
| X79201.1 | - | 5 | 603 | |||

SS18
Genomic Map