siRNA Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005981.3 | NP_005972.1 | 4 | 564 | ||
| XM_005269074.3 | XP_005269131.2 | 4 | 1489 | |||
| XM_017019793.1 | XP_016875282.1 | 2 | 293 | |||
| GenBank mRNA | AK090502.1 | - | 2 | 967 | ||
| AK294128.1 | - | 4 | 517 | |||
| AK315765.1 | - | 4 | 685 | |||
| BC010377.1 | - | 4 | 517 | 4294627 | ||
| CR407647.1 | - | 4 | 410 | |||
| U01160.1 | - | 4 | 512 | |||

TSPAN31
Genomic Map