siRNA Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005981.3 | NP_005972.1 | 4 | 572 | ||
| XM_005269074.3 | XP_005269131.2 | 4 | 1497 | |||
| XM_017019793.1 | XP_016875282.1 | 2 | 301 | |||
| GenBank mRNA | AK090502.1 | - | 2 | 975 | ||
| AK294128.1 | - | 4 | 525 | |||
| AK315765.1 | - | 4 | 693 | |||
| BC010377.1 | - | 4 | 525 | 4294627 | ||
| CR407647.1 | - | 4 | 418 | |||
| U01160.1 | - | 4 | 520 | |||

TSPAN31
Genomic Map