siRNA Details
Gene Symbol: PSKH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006742.2 | NP_006733.1 | 2 | 341 | ||
GenBank mRNA | AJ272212.1 | - | 2 | 301 | ||
AK309272.1 | - | 2 | 341 | |||
BC062616.1 | - | 2 | 312 | 5188431 | ||
DA429976.1 | - | 2 | 341 |
