siRNA Details
| Gene Symbol: RRAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128850.1 | NP_001122322.1 | 5 | 1368 | ||
| NM_004165.2 | NP_004156.1 | 5 | 1359 | |||
| GenBank mRNA | BC057815.1 | - | 5 | 1367 | 30341066 | |
| L24564.1 | - | 5 | 1339 | |||

RRAD
Genomic Map