siRNA Details
| Gene Symbol: PMEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200054.1 | NP_001186983.1 | 3 | 362 | ||
| NM_001320121.1 | NP_001307050.1 | 3 | 362 | |||
| NM_001320122.1 | NP_001307051.1 | 3 | 362 | |||
| NM_006928.4 | NP_008859.1 | 4 | 397 | |||
| XM_006719569.1 | XP_006719632.1 | 3 | 974 | |||
| XM_011538685.1 | XP_011536987.1 | 4 | 426 | |||
| GenBank mRNA | AK092881.1 | - | 3 | 975 | ||
| AK130180.1 | - | 4 | 421 | |||
| BC001414.2 | - | 3 | 321 | 3139788 | ||
| BT007202.1 | - | 3 | 311 | |||
| DA035117.1 | - | 4 | 394 | |||
| DA633173.1 | - | 4 | 388 | |||
| M32295.1 | - | 3 | 332 | |||
| M77348.1 | - | 3 | 322 | |||
| S73003.1 | - | 3 | 332 | |||
| U01874.1 | - | 3 | 317 | |||

PMEL
Genomic Map