siRNA Details
| Gene Symbol: RTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021136.2 | NP_066959.1 | 9 | 3045 | ||
| NM_206852.2 | NP_996734.1 | 7 | 1440 | |||
| XM_017021578.1 | XP_016877067.1 | 7 | 1329 | |||
| GenBank mRNA | AK128113.1 | - | 4 | 2905 | ||
| BC090862.1 | - | 9 | 2960 | 6179803 | ||
| L10333.1 | - | 9 | 2958 | |||
| L10334.1 | - | 7 | 1866 | |||
| L10335.1 | - | 7 | 1160 | |||

RTN1
Genomic Map