siRNA Details
| Gene Symbol: TMSB4Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004202.2 | NP_004193.1 | 2 | 893 | ||
| GenBank mRNA | AF000989.1 | - | 2 | 891 | ||
| BC022482.1 | - | 2 | 156 | |||
| BC119763.1 | - | 2 | 651 | 40119344 | ||
| BC119764.1 | - | 2 | 651 | 40119345 | ||

TMSB4Y
Genomic Map