siRNA Details
| Gene Symbol: NDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000266.3 | NP_000257.1 | 3 | 1854 | ||
| GenBank mRNA | BC029901.1 | - | 2 | 1291 | 5179578 | |
| X65724.1 | - | 3 | 1692 | |||
| X65882.1 | - | 3 | 1682 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDP| Gene Symbol: NDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000266.3 | NP_000257.1 | 3 | 1854 | ||
| GenBank mRNA | BC029901.1 | - | 2 | 1291 | 5179578 | |
| X65724.1 | - | 3 | 1692 | |||
| X65882.1 | - | 3 | 1682 | |||