siRNA Details
| Gene Symbol: ORM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000608.2 | NP_000599.1 | 4 | 498 | ||
| GenBank mRNA | AK303760.1 | - | 3 | 830 | ||
| BC015964.1 | - | 4 | 422 | 3934986 | ||
| BC056239.1 | - | 4 | 422 | 6463124 | ||
| CR456930.1 | - | 4 | 382 | |||

ORM2
Genomic Map