siRNA Details
| Gene Symbol: LIMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204426.1 | NP_001191355.1 | 15 | 3126 | ||
| NM_002314.3 | NP_002305.1 | 16 | 3348 | |||
| GenBank mRNA | AK026544.1 | - | 10 | 2384 | ||
| D26309.1 | - | 17 | 3229 | |||

LIMK1
Genomic Map