siRNA Details
| Gene Symbol: MARCKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002356.6 | NP_002347.5 | 2 | 1630 | ||
| GenBank mRNA | AI142997.1 | - | 1 | 316 | ||
| AK074526.1 | - | 2 | 611 | |||
| BC089040.1 | - | 2 | 1475 | 6378349 | ||
| D10522.1 | - | 2 | 1586 | |||
| M68956.1 | - | 2 | 1528 | |||

MARCKS
Genomic Map