siRNA Details
| Gene Symbol: FADS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004265.3 | NP_004256.1 | 1 | 492 | ||
| GenBank mRNA | AF084559.1 | - | 1 | 284 | ||
| AF126799.1 | - | 1 | 147 | |||
| AK074939.1 | - | 1 | 224 | |||
| BC009011.1 | - | 1 | 226 | 4181037 | ||
| DA233832.1 | - | 1 | 491 | |||

FADS2
Genomic Map