siRNA Details
| Gene Symbol: CEACAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002483.6 | NP_002474.4 | 5 | 1165 | ||
| GenBank mRNA | BC005008.1 | - | 5 | 1086 | 3640231 | |
| M18216.1 | - | 5 | 1092 | |||
| M18728.1 | - | 5 | 1096 | |||
| M29541.1 | - | 5 | 1139 | |||

CEACAM6
Genomic Map