siRNA Details
| Gene Symbol: NEK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146099.1 | NP_001139571.1 | 11 | 1284 | ||
| NM_002498.2 | NP_002489.1 | 12 | 1335 | |||
| NM_152720.2 | NP_689933.1 | 12 | 1354 | |||
| NR_027415.2 | 12 | 1334 | ||||
| GenBank mRNA | AB072828.1 | - | 10 | 953 | ||
| AK131359.1 | - | 10 | 2202 | |||
| AK290259.1 | - | 12 | 1067 | |||
| AL833951.1 | - | 11 | 4717 | |||
| AW364762.1 | - | 2 | 65 | |||
| BC019916.1 | - | 12 | 1062 | 5000918 | ||
| DA407862.1 | - | 6 | 489 | |||
| Z29067.1 | - | 10 | 818 | |||

NEK3
Genomic Map