siRNA Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002567.3 | NP_002558.1 | 2 | 489 | ||
| GenBank mRNA | AK226006.1 | - | 2 | 268 | ||
| AK299414.1 | - | 2 | 248 | |||
| AK308056.1 | - | 2 | 247 | |||
| AK311927.1 | - | 2 | 260 | |||
| BC008714.2 | - | 2 | 262 | 2822606 | ||
| BC017396.1 | - | 2 | 283 | 4074533 | ||
| BC018799.1 | - | 2 | 253 | |||
| BC031102.1 | - | 2 | 251 | 5300366 | ||
| BF978793.1 | - | 2 | 331 | |||
| BP347345.1 | - | 2 | 514 | |||
| D16111.1 | - | 2 | 254 | |||
| EU668351.1 | - | 1 | 22 | |||
| EU794643.1 | - | 2 | 253 | |||
| GQ472218.1 | - | 1 | 22 | |||
| HM131449.1 | - | 2 | 245 | |||
| S76773.1 | - | 1 | 22 | |||
| X75252.1 | - | 2 | 273 | |||
| X85033.1 | - | 2 | 166 | |||

PEBP1
Genomic Map