siRNA Details
| Gene Symbol: RAB3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004283.3 | NP_004274.1 | 5 | 770 | ||
| GenBank mRNA | AF081353.1 | - | 5 | 683 | ||
| AF498933.1 | - | 4 | 509 | |||
| AK313538.1 | - | 5 | 770 | |||
| BC016471.1 | - | 5 | 747 | 3861912 | ||

RAB3D
Genomic Map