siRNA Details
| Gene Symbol: GTPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004286.4 | NP_004277.2 | 4 | 826 | ||
| XM_011530537.2 | XP_011528839.1 | 4 | 2088 | |||
| XM_017029099.1 | XP_016884588.1 | 4 | 2080 | |||
| XM_017029100.1 | XP_016884589.1 | 4 | 2050 | |||
| XM_017029101.1 | XP_016884590.1 | 5 | 667 | |||
| XM_017029102.1 | XP_016884591.1 | 3 | 483 | |||
| XM_017029103.1 | XP_016884592.1 | 3 | 581 | |||
| GenBank mRNA | AK315704.1 | - | 3 | 449 | ||
| BC014075.1 | - | 4 | 664 | |||
| CR456501.1 | - | 4 | 649 | |||
| U87964.1 | - | 4 | 570 | |||

GTPBP1
Genomic Map