siRNA Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002667.4 | NP_002658.1 | 2 | 1327 | ||
| GenBank mRNA | BC005269.1 | - | 2 | 1260 | 3934199 | |
| M63603.1 | - | 2 | 1281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLN| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002667.4 | NP_002658.1 | 2 | 1327 | ||
| GenBank mRNA | BC005269.1 | - | 2 | 1260 | 3934199 | |
| M63603.1 | - | 2 | 1281 | |||