siRNA Details
| Gene Symbol: DRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004147.3 | NP_004138.1 | 4 | 527 | ||
| GenBank mRNA | AF078103.1 | - | 4 | 457 | ||
| AJ005940.1 | - | 4 | 461 | |||
| AK098576.1 | - | 4 | 403 | |||
| AK315659.1 | - | 4 | 485 | |||
| BC019285.1 | - | 4 | 466 | 2823263 | ||
| BC020803.1 | - | 4 | 461 | 4250998 | ||
| BT007237.1 | - | 4 | 396 | |||
| CR456488.1 | - | 4 | 438 | |||
| CR542059.1 | - | 4 | 396 | |||

DRG1
Genomic Map