siRNA Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083592.1 | NP_001077061.1 | 3 | 670 | ||
| NM_005012.3 | NP_005003.2 | 3 | 670 | |||
| XM_011541526.1 | XP_011539828.1 | 3 | 269 | |||
| XM_017001376.1 | XP_016856865.1 | 2 | 570 | |||
| XM_017001377.1 | XP_016856866.1 | 5 | 3213 | |||
| GenBank mRNA | AK308088.1 | - | 3 | 676 | ||
| BC080541.1 | - | 3 | 670 | |||
| BC128386.1 | - | 4 | 280 | |||
| M97675.1 | - | 3 | 646 | |||

ROR1
Genomic Map