siRNA Details
| Gene Symbol: MAGEA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005365.4 | NP_005356.1 | 4 | 1669 | ||
| XM_005262334.3 | XP_005262391.1 | 4 | 1691 | |||
| XM_005262335.3 | XP_005262392.1 | 3 | 1598 | |||
| GenBank mRNA | BC002351.2 | - | 4 | 1669 | 2820966 | |
| Gene Symbol: MAGEA9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080790.1 | NP_001074259.1 | 4 | 1676 | ||
| XM_005278192.3 | XP_005278249.1 | 4 | 1691 | |||
| XM_005278193.3 | XP_005278250.1 | 3 | 1598 | |||

MAGEA9
Genomic Map