siRNA Details
| Gene Symbol: LAMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005560.4 | NP_005551.3 | 37, 38 | 5010 | ||
| XM_006723796.3 | XP_006723859.1 | 37, 38 | 5013 | |||
| XM_006723798.3 | XP_006723861.1 | 37, 38 | 5016 | |||
| XM_011528818.2 | XP_011527120.1 | 36, 37 | 4875 | |||
| XM_011528819.2 | XP_011527121.1 | 35, 36 | 4740 | |||
| GenBank mRNA | AB067494.1 | - | 34, 35 | 4354 | ||
| AF443072.1 | - | 37, 38 | 5010 | |||

LAMA5
Genomic Map