siRNA Details
| Gene Symbol: LAMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005560.4 | NP_005551.3 | 59 | 7952 | ||
| XM_006723796.3 | XP_006723859.1 | 59 | 7955 | |||
| XM_011528818.2 | XP_011527120.1 | 58 | 7817 | |||
| XM_011528819.2 | XP_011527121.1 | 57 | 7682 | |||
| GenBank mRNA | AB011105.1 | - | 14 | 1737 | ||
| AF443072.1 | - | 59 | 7952 | |||

LAMA5
Genomic Map