siRNA Details
| Gene Symbol: LCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005565.3 | NP_005556.1 | 10, 11 | 975 | ||
| GenBank mRNA | AK292890.1 | - | 10, 11 | 936 | ||
| AK296098.1 | - | 2, 3 | 332 | |||
| AK300000.1 | - | 8, 9 | 819 | |||
| AK310369.1 | - | 7, 8 | 586 | |||
| BC016618.1 | - | 10, 11 | 912 | 4476274 | ||
| BT007273.1 | - | 10, 11 | 768 | |||
| FN432830.1 | - | 11, 12 | 848 | |||
| U20158.1 | - | 10, 11 | 1023 | |||

LCP2
Genomic Map