siRNA Details
| Gene Symbol: CD81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297649.1 | NP_001284578.1 | 4, 5 | 471 | ||
| NM_004356.3 | NP_004347.1 | 4, 5 | 589 | |||
| XM_011520492.2 | XP_011518794.1 | 4, 5 | 823 | |||
| XM_017018598.1 | XP_016874087.1 | 4, 5 | 332 | |||
| GenBank mRNA | BC002978.1 | - | 4, 5 | 396 | 3510634 | |
| BC093047.1 | - | 4, 5 | 587 | 30398483 | ||
| BG397506.1 | - | 5, 6 | 459 | |||
| BP323362.1 | - | 4, 5 | 485 | |||
| BQ709484.1 | - | 586 | ||||
| BT019507.1 | - | 4, 5 | 356 | |||
| BT019508.1 | - | 4, 5 | 356 | |||
| M33680.1 | - | 4, 5 | 594 | |||

CD81
Genomic Map