siRNA Details
| Gene Symbol: CAPRIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005898.4 | NP_005889.3 | 10, 11 | 1319 | ||
| NM_203364.2 | NP_976240.1 | 10, 11 | 1319 | |||
| XM_017017749.1 | XP_016873238.1 | 9, 10 | 1290 | |||
| GenBank mRNA | AK127972.1 | - | 9, 10 | 1119 | ||
| BC001731.1 | - | 10, 11 | 1319 | 3355481 | ||
| Z48042.1 | - | 9, 10 | 1172 | |||

CAPRIN1
Genomic Map