siRNA Details
| Gene Symbol: MFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242336.1 | NP_001229265.1 | 2 | 627 | ||
| NM_005927.4 | NP_005918.1 | 2 | 330 | |||
| GenBank mRNA | AK000358.1 | - | 1 | 290 | ||
| AK315573.1 | - | 2 | 299 | |||
| BC026244.1 | - | 2 | 587 | 4697443 | ||
| CB994356.1 | - | 2 | 331 | |||

MFAP3
Genomic Map