siRNA Details
| Gene Symbol: MFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135037.1 | NP_001128509.1 | 2 | 414 | ||
| NM_001242336.1 | NP_001229265.1 | 3 | 1172 | |||
| NM_005927.4 | NP_005918.1 | 3 | 875 | |||
| GenBank mRNA | AK000358.1 | - | 2 | 835 | ||
| AK296468.1 | - | 2 | 414 | |||
| AK315573.1 | - | 3 | 844 | |||
| BC026244.1 | - | 3 | 1132 | 4697443 | ||

MFAP3
Genomic Map