siRNA Details
| Gene Symbol: CTAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001247988.1 | NP_001234917.1 | 23 | 2934 | ||
| NM_001247989.1 | NP_001234918.1 | 24 | 3247 | |||
| NM_001247990.1 | NP_001234919.1 | 24 | 3137 | |||
| NM_005930.3 | NP_005921.2 | 24 | 3232 | |||
| NM_203354.2 | NP_976229.1 | 24 | 2983 | |||
| NM_203355.2 | NP_976230.1 | 23 | 3103 | |||
| NM_203356.2 | NP_976231.1 | 24 | 3063 | |||
| GenBank mRNA | CD102996.1 | - | 1 | 388 | ||
| U94780.1 | - | 24 | 3214 | |||

CTAGE5
Genomic Map