siRNA Details
| Gene Symbol: CTAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001247988.1 | NP_001234917.1 | 23 | 3263 | ||
| NM_001247989.1 | NP_001234918.1 | 24 | 3576 | |||
| NM_001247990.1 | NP_001234919.1 | 24 | 3466 | |||
| NM_005930.3 | NP_005921.2 | 24 | 3561 | |||
| NM_203354.2 | NP_976229.1 | 24 | 3312 | |||
| NM_203355.2 | NP_976230.1 | 23 | 3432 | |||
| NM_203356.2 | NP_976231.1 | 24 | 3392 | |||
| GenBank mRNA | U94780.1 | - | 24 | 3541 | ||

CTAGE5
Genomic Map