siRNA Details
| Gene Symbol: ASMTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173473.1 | NP_001166944.1 | 6 | 498 | ||
| NM_001173474.1 | NP_001166945.1 | 5 | 535 | |||
| NM_004192.3 | NP_004183.2 | 6 | 583 | |||
| XM_005274434.3 | XP_005274491.1 | 7 | 689 | |||
| XM_011546185.2 | XP_011544487.1 | 4 | 331 | |||
| XM_017029919.1 | XP_016885408.1 | 5 | 379 | |||
| GenBank mRNA | AK090498.1 | - | 5 | 487 | ||
| AK225770.1 | - | 6 | 505 | |||
| AK297805.1 | - | 5 | 416 | |||
| AK299231.1 | - | 6 | 501 | |||
| AK301844.1 | - | 6 | 492 | |||
| BC002508.2 | - | 6 | 469 | 3051194 | ||
| BC010089.2 | - | 6 | 467 | 2988425 | ||
| BM129217.1 | - | 1 | 32 | |||
| DA301876.1 | - | 6 | 538 | |||

ASMTL
Genomic Map