siRNA Details
| Gene Symbol: MTNR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005959.3 | NP_005950.1 | 2 | 1611 | ||
| GenBank mRNA | BC069163.1 | - | 2 | 1611 | 7216982 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTNR1B| Gene Symbol: MTNR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005959.3 | NP_005950.1 | 2 | 1611 | ||
| GenBank mRNA | BC069163.1 | - | 2 | 1611 | 7216982 | |
Set Membership: |
|