siRNA Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126181.1 | NP_001119653.1 | 4 | 783 | ||
| NM_006176.2 | NP_006167.1 | 4 | 780 | |||
| GenBank mRNA | BC002835.2 | - | 4 | 671 | 3660038 | |
| U89165.1 | - | 4 | 768 | |||
| Y09689.1 | - | 4 | 757 | |||

NRGN
Genomic Map