siRNA Details
| Gene Symbol: LETM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012318.2 | NP_036450.1 | 14 | 3626 | ||
| XM_006713884.1 | XP_006713947.1 | 14 | 3331 | |||
| XM_017008187.1 | XP_016863676.1 | 13 | 3747 | |||
| GenBank mRNA | AF061025.1 | - | 14 | 3626 | ||
| BC021208.1 | - | 14 | 3551 | 4126510 | ||

LETM1
Genomic Map