siRNA Details
| Gene Symbol: MKLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145354.1 | NP_001138826.1 | 7 | 670 | ||
| NM_001321316.1 | NP_001308245.1 | 6 | 601 | |||
| NM_013255.4 | NP_037387.2 | 6 | 598 | |||
| XM_006715993.2 | XP_006716056.1 | 6 | 598 | |||
| XM_011516224.2 | XP_011514526.1 | 6 | 598 | |||
| GenBank mRNA | AF047489.1 | - | 6 | 552 | ||
| AK002024.1 | - | 6 | 540 | |||
| AK294384.1 | - | 7 | 671 | |||
| AK310452.1 | - | 6 | 551 | |||
| AL705061.1 | - | 6 | 542 | |||
| BC067825.1 | - | 6 | 555 | 5268939 | ||

MKLN1
Genomic Map