siRNA Details
| Gene Symbol: OGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014057.4 | NP_054776.1 | 4 | 651 | ||
| NM_024416.4 | NP_077727.3 | 4 | 570 | |||
| NM_033014.3 | NP_148935.1 | 4 | 933 | |||
| GenBank mRNA | AF086912.1 | - | 4 | 498 | ||
| AF100758.1 | - | 4 | 498 | |||
| AF202167.1 | - | 3 | 423 | |||
| AK075521.1 | - | 3 | 462 | |||
| AK129845.1 | - | 3 | 455 | |||
| AK289628.1 | - | 4 | 648 | |||
| AK295433.1 | - | 4 | 570 | |||
| AL110267.1 | - | 3 | 440 | |||
| BC037273.2 | - | 3 | 439 | 5265880 | ||
| BC095443.1 | - | 3 | 455 | 30405707 | ||
| BG772245.1 | - | 4 | 574 | |||
| BX646984.1 | - | 4 | 608 | |||
| CR533516.1 | - | 3 | 372 | |||

OGN
Genomic Map