siRNA Details
| Gene Symbol: TM4SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014220.2 | NP_055035.1 | 5 | 872 | ||
| XM_017006385.1 | XP_016861874.1 | 5 | 892 | |||
| GenBank mRNA | AK093829.1 | - | 2 | 2300 | ||
| AK130073.1 | - | 5 | 784 | |||
| AK130271.1 | - | 5 | 866 | |||
| AL832780.1 | - | 1 | 6347 | |||
| BC008442.1 | - | 5 | 739 | 4101110 | ||
| BC010166.2 | - | 5 | 688 | 4040623 | ||
| BC034145.1 | - | 5 | 832 | 4618019 | ||
| M90657.1 | - | 746 | ||||

TM4SF1
Genomic Map