siRNA Details
| Gene Symbol: ZNF235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004234.4 | NP_004225.3 | 4 | 318 | ||
| XM_005259412.4 | XP_005259469.1 | 4 | 4386 | |||
| GenBank mRNA | AK300317.1 | - | 4 | 318 | ||
| AK307103.1 | - | 4 | 312 | |||
| AK315242.1 | - | 4 | 293 | |||
| BC002663.2 | - | 3 | 264 | |||
| BC002800.2 | - | 3 | 264 | |||
| BC105807.1 | - | 4 | 297 | |||
| BC117215.1 | - | 4 | 303 | 40125766 | ||
| BC143482.1 | - | 4 | 303 | |||

ZNF235
Genomic Map