siRNA Details
| Gene Symbol: ABCB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000443.3 | NP_000434.1 | 20 | 2499 | ||
| NM_018849.2 | NP_061337.1 | 20 | 2499 | |||
| NM_018850.2 | NP_061338.1 | 20 | 2499 | |||
| XM_011516308.2 | XP_011514610.2 | 20 | 2523 | |||
| XM_011516309.2 | XP_011514611.2 | 20 | 2523 | |||
| XM_011516310.2 | XP_011514612.2 | 19 | 2418 | |||
| XM_011516311.2 | XP_011514613.2 | 20 | 2523 | |||
| XM_011516312.2 | XP_011514614.2 | 20 | 2523 | |||
| XM_011516313.2 | XP_011514615.1 | 20 | 2522 | |||
| XM_011516315.2 | XP_011514617.2 | 15 | 1899 | |||
| XM_017012323.1 | XP_016867812.1 | 20 | 2510 | |||
| GenBank mRNA | BC042531.1 | - | 20 | 2535 | 4837724 | |
| M23234.1 | - | 20 | 2455 | |||

ABCB4
Genomic Map