siRNA Details
| Gene Symbol: OSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319108.1 | NP_001306037.1 | 2 | 365 | ||
| NM_020530.5 | NP_065391.1 | 2 | 184 | |||
| GenBank mRNA | AB451493.1 | - | 1 | 65 | ||
| BC011589.1 | - | 2 | 181 | 4548943 | ||
| BM562358.1 | - | 2 | 377 | |||
| CR456534.1 | - | 2 | 154 | |||
| CR541703.1 | - | 2 | 140 | |||
| CR981101.1 | - | 2 | 184 | |||

OSM
Genomic Map